Gene Transcripts

Gene Symbol
NHSL2
Entrez Gene ID
340527
Gene Name
NHS like 2
Gene Aliases
Chromosome Location
Chr. X: 71911073 - 72153286 on Build GRCh38
UniGene ID
Hs.397836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011530934.2 XP_011529236.1 8 15794
XM_017029473.1 XP_016884962.1 8 14764
XM_017029474.1 XP_016884963.1 8 10906
XM_017029475.1 XP_016884964.1 4 14149
XM_017029476.1 XP_016884965.1 4 10266
XM_017029477.1 XP_016884966.1 4 15180
GenBank mRNA AK056217.1 - 1 15

View Full Product Details