Gene Transcripts
- Gene Symbol
- CARS
- Entrez Gene ID
- 833
- Gene Name
- cysteinyl-tRNA synthetase
- Gene Aliases
- CARS1, CYSRS, MGC:11246
- Chromosome Location
- Chr. 11: 3000922 - 3057451 on Build GRCh38
- UniGene ID
- Hs.274873
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK131221.1 |
- |
1 |
1034 |
|
View Full Product Details