Gene Transcripts
- Gene Symbol
- USP34
- Entrez Gene ID
- 9736
- Gene Name
- ubiquitin specific peptidase 34
- Gene Aliases
- Chromosome Location
- Chr. 2: 61187455 - 61470714 on Build GRCh38
- UniGene ID
- Hs.644708
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK024341.1 |
- |
1 |
1767 |
|
View Full Product Details