Gene Transcripts

Gene Symbol
BACE2
Entrez Gene ID
25825
Gene Name
beta-site APP-cleaving enzyme 2
Gene Aliases
AEPLC, ALP56, ASP1, ASP21, BAE2, CDA13, CEAP1, DRAP
Chromosome Location
Chr. 21: 41167801 - 41282534 on Build GRCh38
UniGene ID
Hs.43047
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012105.4 NP_036237.2 9 6100
NM_138991.2 NP_620476.1 8 5950
NM_138992.2 NP_620477.1 8 5931
GenBank mRNA AK023647.1 - 1 606
BC020582.1 - 1 482

View Full Product Details