Gene Transcripts
- Gene Symbol
- LOC642648
- Entrez Gene ID
- 642648
- Gene Name
- translation initiation factor IF-2
- Gene Aliases
- Chromosome Location
- Chr. 22: 46067464 - 46071470 on Build GRCh38
- UniGene ID
- Hs.631363
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123383.1 |
- |
1 |
2216 |
|
View Full Product Details