Gene Transcripts
- Gene Symbol
- KIRREL
- Entrez Gene ID
- 55243
- Gene Name
- kin of IRRE like (Drosophila)
- Gene Aliases
- NEPH1
- Chromosome Location
- Chr. 1: 157993273 - 158100262 on Build GRCh38
- UniGene ID
- Hs.272234
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA028774.1 |
- |
1 |
197 |
|
Chr. 1 - 158097908 - 158098469 [+] |
GenBankmRNA |
BU196665.1 |
- |
|
284 |
|
Chr. - - [-] |
GenBankmRNA |
BQ677239.1 |
- |
1 |
284 |
|
Chr. 1 - 158097820 - 158098704 [+] |
GenBankmRNA |
CV815855.1 |
- |
1 |
115 |
|
Chr. 1 - 158097989 - 158098866 [+] |
View Full Product Details