Gene Transcripts
- Gene Symbol
- ARPC4
- Entrez Gene ID
- 10093
- Gene Name
- actin related protein 2/3 complex subunit 4
- Gene Aliases
- ARC20, P20-ARC
- Chromosome Location
- Chr. 3: 9792495 - 9807105 on Build GRCh38
- UniGene ID
- Hs.323342
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK074205.1 |
- |
1 |
147 |
|
View Full Product Details