Gene Transcripts
- Gene Symbol
- PACRGL
- Entrez Gene ID
- 133015
- Gene Name
- PARK2 coregulated like
- Gene Aliases
- C4orf28
- Chromosome Location
- Chr. 4: 20696282 - 20757766 on Build GRCh38
- UniGene ID
- Hs.479298
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ718422.1 |
- |
1 |
785 |
|
Chr. 4 - 20731176 - 20732094 [+] |
GenBankmRNA |
AK125392.1 |
- |
2 |
2093 |
|
Chr. 4 - 20729805 - 20732554 [+] |
View Full Product Details