Gene Transcripts
- Gene Symbol
- ENTPD1
- Entrez Gene ID
- 953
- Gene Name
- ectonucleoside triphosphate diphosphohydrolase 1
- Gene Aliases
- ATPDase, CD39, NTPDase-1, SPG64
- Chromosome Location
- Chr. 10: 95710901 - 95877266 on Build GRCh38
- UniGene ID
- Hs.576612
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098278.1 |
- |
1 |
1489 |
|
View Full Product Details