Gene Transcripts
- Gene Symbol
- RAB7A
- Entrez Gene ID
- 7879
- Gene Name
- RAB7A, member RAS oncogene family
- Gene Aliases
- PRO2706, RAB7
- Chromosome Location
- Chr. 3: 128726136 - 128814798 on Build GRCh38
- UniGene ID
- Hs.684374
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC014200.2 |
- |
1 |
1585 |
|
View Full Product Details