Gene Transcripts
- Gene Symbol
- RPAIN
- Entrez Gene ID
- 84268
- Gene Name
- RPA interacting protein
- Gene Aliases
- HRIP, RIP
- Chromosome Location
- Chr. 17: 5419185 - 5433020 on Build GRCh38
- UniGene ID
- Hs.462086
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098491.1 |
- |
2 |
648 |
|
View Full Product Details