Gene Transcripts

Gene Symbol
FAM87A
Entrez Gene ID
157693
Gene Name
family with sequence similarity 87 member A
Gene Aliases
Chromosome Location
Chr. 8: 375931 - 383174 on Build GRCh38
UniGene ID
Hs.591390
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_103537.1 3 1876
GenBank mRNA BC037297.2 - 3 1876


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000330148.6 - 3 1876 Chr. 8 - 375934 - 383174 [-]

View Full Product Details