Gene Transcripts
- Gene Symbol
- PRKG1-AS1
- Entrez Gene ID
- 100506939
- Gene Name
- PRKG1 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 10: 52296848 - 52314128 on Build GRCh38
- UniGene ID
- Hs.584791
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC039504.2 |
- |
4 |
371 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM479024.1 |
- |
4 |
384 |
|
Chr. 10 - 52252147 - 52297917 [-] |
View Full Product Details