Gene Transcripts

Gene Symbol
SEPT7
Entrez Gene ID
989
Gene Name
septin 7
Gene Aliases
CDC10, CDC3, NBLA02942, SEPT7A
Chromosome Location
Chr. 7: 35800986 - 35915763 on Build GRCh38
UniGene ID
Hs.191346
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011553.3 NP_001011553.2 13 4283
NM_001242956.1 NP_001229885.1 12 4178
NM_001788.5 NP_001779.3 14 4286


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CR616642.1 - 1 788 Chr. 7 - 35906254 - 35907087 [+]
GenBankmRNA AL521817.3 - 1 787 Chr. 7 - 35906254 - 35907087 [+]
GenBankmRNA H52870.1 - 1 1768 Chr. 7 - 35905266 - 35907105 [+]

View Full Product Details