Gene Transcripts
- Gene Symbol
- SEPT7
- Entrez Gene ID
- 989
- Gene Name
- septin 7
- Gene Aliases
- CDC10, CDC3, NBLA02942, SEPT7A
- Chromosome Location
- Chr. 7: 35800986 - 35915763 on Build GRCh38
- UniGene ID
- Hs.191346
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CR616642.1 |
- |
1 |
663 |
|
Chr. 7 - 35906254 - 35907087 [+] |
GenBankmRNA |
AL521817.3 |
- |
1 |
663 |
|
Chr. 7 - 35906254 - 35907087 [+] |
GenBankmRNA |
H52870.1 |
- |
1 |
1643 |
|
Chr. 7 - 35905266 - 35907105 [+] |
View Full Product Details