Gene Transcripts

Gene Symbol
SPINK13
Entrez Gene ID
153218
Gene Name
serine peptidase inhibitor, Kazal type 13 (putative)
Gene Aliases
HBVDNAPTP1, HESPINTOR, LiESP6, SPINK5L3
Chromosome Location
Chr. 5: 148268860 - 148286210 on Build GRCh38
UniGene ID
Hs.483771
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK001520.1 - 1 113


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AU129688.1 - 1 113 Chr. 5 - 148268180 - 148282204 [+]

View Full Product Details