Gene Transcripts
- Gene Symbol
- TPD52L1
- Entrez Gene ID
- 7164
- Gene Name
- tumor protein D52-like 1
- Gene Aliases
- D53
- Chromosome Location
- Chr. 6: 125153483 - 125264407 on Build GRCh38
- UniGene ID
- Hs.591347
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK090516.1 |
- |
1 |
1720 |
|
View Full Product Details