Gene Transcripts
- Gene Symbol
- FEZ2
- Entrez Gene ID
- 9637
- Gene Name
- fasciculation and elongation protein zeta 2
- Gene Aliases
- HUM3CL
- Chromosome Location
- Chr. 2: 36552239 - 36598190 on Build GRCh38
- UniGene ID
- Hs.258563
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK057492.1 |
- |
1 |
490 |
|
View Full Product Details