Gene Transcripts
- Gene Symbol
- STAM-AS1
- Entrez Gene ID
- 102723166
- Gene Name
- STAM antisense RNA 1 (head to head)
- Gene Aliases
- locus3182
- Chromosome Location
- Chr. 10: 17641284 - 17643918 on Build GRCh38
- UniGene ID
- Hs.547311
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110370.1 |
|
1 |
1430 |
|
GenBank mRNA |
AK123677.1 |
- |
1 |
1391 |
|
View Full Product Details