Gene Transcripts

Gene Symbol
NCBP3
Entrez Gene ID
55421
Gene Name
nuclear cap binding subunit 3
Gene Aliases
C17orf85, ELG, HSA277841
Chromosome Location
Chr. 17: 3806748 - 3846251 on Build GRCh38
UniGene ID
Hs.120963
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114118.2 NP_001107590.1 13 7861
XM_005256717.2 XP_005256774.1 14 7399
GenBank mRNA AK056724.1 - 1 1781

View Full Product Details