Gene Transcripts
- Gene Symbol
- DDX3X
- Entrez Gene ID
- 1654
- Gene Name
- DEAD-box helicase 3, X-linked
- Gene Aliases
- CAP-Rf, DBX, DDX14, DDX3, HLP2, MRX102
- Chromosome Location
- Chr. X: 41333308 - 41364472 on Build GRCh38
- UniGene ID
- Hs.728563
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK127902.1 |
- |
1 |
1520 |
|
- Gene Symbol
- LOC107985678
- Entrez Gene ID
- 107985678
- Gene Name
- uncharacterized LOC107985678
- Gene Aliases
- Chromosome Location
- Chr. X: 41334304 - 41337401 on Build GRCh38
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CK005511.1 |
- |
1 |
150 |
|
Chr. X - 41335980 - 41336636 [+] |
View Full Product Details