Gene Transcripts
- Gene Symbol
- TMTC4
- Entrez Gene ID
- 84899
- Gene Name
- transmembrane and tetratricopeptide repeat containing 4
- Gene Aliases
- Chromosome Location
- Chr. 13: 100603836 - 100675025 on Build GRCh38
- UniGene ID
- Hs.190983
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124599.1 |
- |
1 |
2330 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB269704.1 |
- |
1 |
170 |
|
Chr. 13 - 100605411 - 100605978 [-] |
View Full Product Details