Gene Transcripts
- Gene Symbol
- PPIE
- Entrez Gene ID
- 10450
- Gene Name
- peptidylprolyl isomerase E
- Gene Aliases
- CYP-33, CYP33
- Chromosome Location
- Chr. 1: 39738845 - 39765698 on Build GRCh38
- UniGene ID
- Hs.524690
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ187153.1 |
- |
1 |
180 |
|
Chr. 1 - 39756172 - 39756771 [+] |
GenBankmRNA |
AA648586.1 |
- |
1 |
48 |
|
Chr. 1 - 39756330 - 39756678 [+] |
View Full Product Details