Gene Transcripts

Gene Symbol
PDK3
Entrez Gene ID
5165
Gene Name
pyruvate dehydrogenase kinase 3
Gene Aliases
CMTX6, GS1-358P8.4
Chromosome Location
Chr. X: 24465227 - 24550466 on Build GRCh38
UniGene ID
Hs.296031
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142386.2 NP_001135858.1 12 10850
GenBank mRNA BC038512.2 - 1 2652

View Full Product Details