Gene Transcripts
- Gene Symbol
- FAM204A
- Entrez Gene ID
- 63877
- Gene Name
- family with sequence similarity 204 member A
- Gene Aliases
- C10orf84, bA319I23.1
- Chromosome Location
- Chr. 10: 118305889 - 118342352 on Build GRCh38
- UniGene ID
- Hs.105575
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX538256.1 |
- |
1 |
266 |
|
View Full Product Details