Gene Transcripts

Gene Symbol
SIGMAR1
Entrez Gene ID
10280
Gene Name
sigma non-opioid intracellular receptor 1
Gene Aliases
ALS16, DSMA2, OPRS1, SIG-1R, SR-BP, SR-BP1, SRBP, hSigmaR1, sigma1R
Chromosome Location
Chr. 9: 34634722 - 34637826 on Build GRCh38
UniGene ID
Hs.522087
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282208.1 NP_001269137.1 3 493
GenBank mRNA BC006307.1 - 686
BI596428.1 - 3 449

View Full Product Details