Gene Transcripts

Gene Symbol
SHISA6
Entrez Gene ID
388336
Gene Name
shisa family member 6
Gene Aliases
Chromosome Location
Chr. 17: 11241423 - 11564063 on Build GRCh38
UniGene ID
Hs.592124
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173461.1 NP_001166932.1 4 6150
NM_001173462.1 NP_001166933.1 5 6246
NM_207386.3 NP_997269.2 6 6303
XM_017024618.1 XP_016880107.1 5 6207
GenBank mRNA AK094349.1 - 1 1194
AK095052.1 - 1 1796
BC105704.1 - 1 952
BC105705.1 - 1 952
BC105706.1 - 1 951
BC105707.1 - 1 952

View Full Product Details