Gene Transcripts
- Gene Symbol
- CENPL
- Entrez Gene ID
- 91687
- Gene Name
- centromere protein L
- Gene Aliases
- C1orf155, CENP-L, dJ383J4.3
- Chromosome Location
- Chr. 1: 173799550 - 173824639 on Build GRCh38
- UniGene ID
- Hs.531856
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK056348.1 |
- |
1 |
1543 |
|
View Full Product Details