Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001270396.1 | NP_001257325.1 | 6 | 2274 | |
NR_072995.1 | 7 | 2404 | |||
XM_011537949.2 | XP_011536251.1 | 6 | 2188 | ||
XM_011537950.2 | XP_011536252.1 | 6 | 2160 | ||
XM_017018848.1 | XP_016874337.1 | 7 | 2264 | ||
XM_017018849.1 | XP_016874338.1 | 7 | 2290 | ||
GenBank mRNA | AA649108.1 | - | 1 | 222 | |
BC032022.1 | - | 1 | 885 |