Gene Transcripts
- Gene Symbol
- LOC105372526
- Entrez Gene ID
- 105372526
- Gene Name
- uncharacterized LOC105372526
- Gene Aliases
- Chromosome Location
- Chr. 20: 10659560 - 10663956 on Build GRCh38
- UniGene ID
- Hs.664262
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK023793.1 |
- |
1 |
2585 |
|
- Gene Symbol
- JAG1
- Entrez Gene ID
- 182
- Gene Name
- jagged 1
- Gene Aliases
- AGS, AHD, AWS, CD339, HJ1, JAGL1
- Chromosome Location
- Chr. 20: 10637684 - 10674046 on Build GRCh38
- UniGene ID
- Hs.224012
- Species
- Homo sapiens
View Full Product Details