Gene Transcripts
- Gene Symbol
- KXD1
- Entrez Gene ID
- 79036
- Gene Name
- KxDL motif containing 1
- Gene Aliases
- BORCS4, C19orf50, KXDL, MST096, MSTP096
- Chromosome Location
- Chr. 19: 18557762 - 18569387 on Build GRCh38
- UniGene ID
- Hs.714548
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX648739.1 |
- |
5 |
2449 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
HY336111.1 |
- |
1 |
261 |
|
Chr. 19 - 18559118 - 18559548 [-] |
View Full Product Details