Gene Transcripts

Gene Symbol
HIVEP3
Entrez Gene ID
59269
Gene Name
human immunodeficiency virus type I enhancer binding protein 3
Gene Aliases
KBP-1, KBP1, KRC, SHN3, Schnurri-3, ZAS3, ZNF40C
Chromosome Location
Chr. 1: 41506365 - 42035925 on Build GRCh38
UniGene ID
Hs.403972
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_038260.1 3 1028
NR_038261.1 4 846
GenBank mRNA BC015242.1 - 1 109
BC040942.1 - 3 872
BC046120.1 - 4 846

View Full Product Details