Gene Transcripts

Gene Symbol
MEAF6
Entrez Gene ID
64769
Gene Name
MYST/Esa1 associated factor 6
Gene Aliases
C1orf149, CENP-28, EAF6, NY-SAR-91
Chromosome Location
Chr. 1: 37489960 - 37514819 on Build GRCh38
UniGene ID
Hs.17118
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270875.1 NP_001257804.1 7 4745
NM_001270876.1 NP_001257805.1 6 4418
NM_022756.5 NP_073593.2 8 4775
NR_073090.1 9 4833
NR_073091.1 8 4803
NR_073092.1 9 4864


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AF172817.1 - 1 124 Chr. 1 - 37487118 - 37490126 [-]

View Full Product Details