Gene Transcripts

Gene Symbol
GHR
Entrez Gene ID
2690
Gene Name
growth hormone receptor
Gene Aliases
GHBP, GHIP
Chromosome Location
Chr. 5: 42423775 - 42721878 on Build GRCh38
UniGene ID
Hs.125180
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242404.2 NP_001229333.1 1, 2 138
GenBank mRNA Z11849.1 - 138

View Full Product Details