Gene Transcripts
- Gene Symbol
- NPAS2
- Entrez Gene ID
- 4862
- Gene Name
- neuronal PAS domain protein 2
- Gene Aliases
- MOP4, PASD4, bHLHe9
- Chromosome Location
- Chr. 2: 100820151 - 100996829 on Build GRCh38
- UniGene ID
- Hs.156832
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC035412.1 |
- |
1 |
1055 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000452364.1 |
- |
1 |
1055 |
|
Chr. 2 - 100993676 - 101002244 [-] |
View Full Product Details