Gene Transcripts

Gene Symbol
EBF2
Entrez Gene ID
64641
Gene Name
early B-cell factor 2
Gene Aliases
COE2, EBF-2, O/E-3, OE-3
Chromosome Location
Chr. 8: 25841730 - 26045124 on Build GRCh38
UniGene ID
Hs.491292
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022659.3 NP_073150.2 16 3070
GenBank mRNA AK093878.1 - 1 395
BQ878633.1 - 1 222

View Full Product Details