Gene Transcripts

Gene Symbol
COMMD5
Entrez Gene ID
28991
Gene Name
COMM domain containing 5
Gene Aliases
HCARG, HT002
Chromosome Location
Chr. 8: 144837978 - 144853578 on Build GRCh38
UniGene ID
Hs.493218
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK092477.1 - 2 2153

View Full Product Details