Gene Transcripts
- Gene Symbol
- ACAD8
- Entrez Gene ID
- 27034
- Gene Name
- acyl-CoA dehydrogenase family member 8
- Gene Aliases
- ACAD-8, ARC42
- Chromosome Location
- Chr. 11: 134253532 - 134265858 on Build GRCh38
- UniGene ID
- Hs.14791
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124235.1 |
- |
1 |
1195 |
|
View Full Product Details