Gene Transcripts

Gene Symbol
HSPE1P13
Entrez Gene ID
106480256
Gene Name
heat shock protein family E (Hsp10) member 1 pseudogene 13
Gene Aliases
Chromosome Location
Chr. 2: 39098149 - 39098445 on Build GRCh38
UniGene ID
Hs.688190
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC047033.1 - 1 602


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BG122370.1 - 1 595 Chr. 2 - 39097991 - 39098686 [-]

View Full Product Details