Gene Transcripts

Gene Symbol
C5orf42
Entrez Gene ID
65250
Gene Name
chromosome 5 open reading frame 42
Gene Aliases
Hug, JBTS17, OFD6
Chromosome Location
Chr. 5: 37063928 - 37249428 on Build GRCh38
UniGene ID
Hs.586199
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011514089.2 XP_011512391.1 53 11180
GenBank mRNA AK022380.1 - 1 1498

View Full Product Details