Gene Transcripts

Gene Symbol
TSL
Entrez Gene ID
790953
Gene Name
testis-expressed, seven-twelve, leukemia
Gene Aliases
Chromosome Location
Chr. 7: 27615974 - 27627492 on Build GRCh38
UniGene ID
Hs.571330
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_122122.1 3 1287
NR_122123.1 2 1067
GenBank mRNA AB050001.1 - 2 520
AB050002.1 - 2 395
AB050003.1 - 3 1286

View Full Product Details