Gene Transcripts
- Gene Symbol
- PIGV
- Entrez Gene ID
- 55650
- Gene Name
- phosphatidylinositol glycan anchor biosynthesis class V
- Gene Aliases
- GPI-MT-II, HPMRS1, PIG-V
- Chromosome Location
- Chr. 1: 26787963 - 26798403 on Build GRCh38
- UniGene ID
- Hs.259605
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK097053.1 |
- |
1 |
514 |
|
View Full Product Details