Gene Transcripts

Gene Symbol
YY1
Entrez Gene ID
7528
Gene Name
YY1 transcription factor
Gene Aliases
DELTA, INO80S, NF-E1, UCRBP, YIN-YANG-1
Chromosome Location
Chr. 14: 100238765 - 100279034 on Build GRCh38
UniGene ID
Hs.388927
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003403.4 NP_003394.1 5 2732
GenBank mRNA AF147416.1 - 1 230
DA234524.1 - 1 161


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA576611.1 - 1 137 Chr. 14 - 100278471 - 100279039 [+]
GenBankmRNA BG678770.1 - 56 Chr. - - [-]
GenBankmRNA BM479654.1 - 1 33 Chr. 14 - 100278575 - 100279577 [+]
GenBankmRNA DA634593.1 - 1 53 Chr. 14 - 100278555 - 100279122 [+]
GenBankmRNA DA695998.1 - 1 212 Chr. 14 - 100278396 - 100278956 [+]
GenBankmRNA DB164168.1 - 1 77 Chr. 14 - 100278531 - 100279103 [+]
GenBankmRNA DA691330.1 - 1 212 Chr. 14 - 100278396 - 100278962 [+]
GenBankmRNA DA819939.1 - 1 150 Chr. 14 - 100278458 - 100279012 [+]
GenBankmRNA DA812758.1 - 1 98 Chr. 14 - 100278510 - 100279232 [+]
GenBankmRNA DA493496.1 - 1 212 Chr. 14 - 100278396 - 100278968 [+]
GenBankmRNA DA507070.1 - 1 211 Chr. 14 - 100278397 - 100278962 [+]

View Full Product Details