Gene Transcripts

Gene Symbol
RFC3
Entrez Gene ID
5983
Gene Name
replication factor C subunit 3
Gene Aliases
RFC38
Chromosome Location
Chr. 13: 33818049 - 33973946 on Build GRCh38
UniGene ID
Hs.115474
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011535174.2 XP_011533476.1 10 4274
GenBank mRNA CR749565.1 - 1 3156

View Full Product Details