Gene Transcripts
- Gene Symbol
- TCEB3
- Entrez Gene ID
- 6924
- Gene Name
- transcription elongation factor B subunit 3
- Gene Aliases
- EloA, SIII, SIII p110, TCEB3A
- Chromosome Location
- Chr. 1: 23743366 - 23762059 on Build GRCh38
- UniGene ID
- Hs.15535
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC019949.1 |
- |
1 |
821 |
|
View Full Product Details