Gene Transcripts
- Gene Symbol
- VN2R9P
- Entrez Gene ID
- 648315
- Gene Name
- vomeronasal 2 receptor 9 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 11: 58040768 - 58066959 on Build GRCh38
- UniGene ID
- Hs.652053
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC067882.1 |
- |
3 |
553 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB990186.1 |
- |
3 |
580 |
|
Chr. 11 - 58047255 - 58060141 [-] |
View Full Product Details