Gene Transcripts
- Gene Symbol
- GLUD1P7
- Entrez Gene ID
- 414212
- Gene Name
- glutamate dehydrogenase 1 pseudogene 7
- Gene Aliases
- GLUD1P2, GLUDP2, GLUDP7, bA342C24.3
- Chromosome Location
- Chr. 10: 46759023 - 46786770 on Build GRCh38
- UniGene ID
- Hs.661155
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_111968.1 |
|
3, 4 |
199 |
|
GenBank mRNA |
BX281697.1 |
- |
3, 4 |
199 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CR604707.1 |
- |
2, 3 |
1226 |
|
Chr. 10 - 46759053 - 46786811 [-] |
View Full Product Details