Gene Transcripts

Gene Symbol
MNT
Entrez Gene ID
4335
Gene Name
MAX network transcriptional repressor
Gene Aliases
MAD6, MXD6, ROX, bHLHd3
Chromosome Location
Chr. 17: 2384060 - 2400964 on Build GRCh38
UniGene ID
Hs.626579
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017024654.1 XP_016880143.1 1 855
GenBank mRNA AF318360.1 - 1 1395
AK001561.1 - 1 856

View Full Product Details