Gene Transcripts
- Gene Symbol
- FRMD6-AS1
- Entrez Gene ID
- 145438
- Gene Name
- FRMD6 antisense RNA 1
- Gene Aliases
- C14orf82
- Chromosome Location
- Chr. 14: 51649516 - 51651744 on Build GRCh38
- UniGene ID
- Hs.645410
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037676.1 |
|
1 |
1652 |
|
GenBank mRNA |
AK022437.1 |
- |
1 |
1653 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
N34909.1 |
- |
1 |
190 |
|
Chr. 14 - 51649983 - 51650282 [-] |
View Full Product Details