Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_017018402.1 | XP_016873891.1 | 4 | 1870 | |
GenBank mRNA | AL050172.1 | - | 1 | 914 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ215272.1 | - | 205 | Chr. - - [-] |